Sterilization Wraps
BHD Sterilization Wraps
Designed for steam sterilization; but also compatible with ETO or gamma irradiation
Heavy Duty SMS Sequential Sterilization Wraps
Strong, rip and tear resistant 50 gsm SMS material
Super Duty SMS Sequential Sterilization Wraps
Strong, rip and tear resistant 88 gsm SMS material
2-Ply SMS, One-Step Heavy Duty Sterilization Wrap
These 2-ply, 50 gsm wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
Manufactured and packaged in a certified ISO 8 Cleanroom.
2-Ply SMS, One-Step Super Duty Sterilization Wrap
These 2-ply, 88 gsm wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
Manufactured and packaged in a certified ISO 8 Cleanroom.
1422A Tyvek® Sterilization Wrap
Drapeable 1422A Tyvek® Wrappers are an improvement over cellulose or SMS based wrapping materials for steam sterilization.
1421B PureClean™ Tyvek® Sterilization Wrap
This new, uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.