DuPont™ ProShield® 30 Shoe Covers
Supplier: DuPont
PE440SWHLG020000 PE440SBUXL020000 PE440SBULG020000 PE440SWHXL020000 PE440SWHMD020000
80076-700CS 123.74 USD
80076-700 80076-705 80076-704 80076-702 76200-886
DuPont™ ProShield® 30 Shoe Covers
Disposable Garments
ProShield® 30 shoe and boot covers offer exceptional skid resistance.
  • Serged seams
  • Elastic opening
  • Skid-resistant
ProShield® 30 is made from spun bonded polypropylene with a polyethylene coating. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
Envase: Bulk packaged garments are bulk packaged in double transparent poly liners.