Pre-Sterilized Disposable Products
Pre-Sterilized Sterility Maintenance and Transport Bags
Prolong the storage and shelf-life of products sterilized ‘in-house’
Sterile Sharpie Markers
Ready to use sterile cleanroom markers eliminate the risks associated with surface sanitization procedures.
Sterile Bic Pens
Ready to use sterile cleanroom pens eliminate the risks associated with surface sanitization procedures.
Sterile Cleanroom Trash Bags
Ready to use pre-sterilized trash bags make it easy to remove waste from pharmaceutical production areas without introducing potential sources of contamination.
Sterile Mop Bucket Liners
Reduce the the number of janitorial steam sterilization loads by using pre-sterilized mop bucket liners.
Extra Long Sterile Tyvek Sleeve Covers
Extra-long sterile Tyvek sleeves are packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitization agents. The sleeves are individually packed and specially folded for easy aseptic donning.