Autoclave Bags
Self-Sealing 1421B Tyvek bag with See Through Film
PureClean™ self-sealing autoclave bags are made from a transparent film and 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.
1422A Tyvek Heat Seal Autoclave Bag
Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
Self-Sealing 1422A Tyvek Autoclave Bag
Self-sealing Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
1422A Tyvek Drawstring Autoclave Bag
The drape-able nature of the Tyvek® 1422A material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
Continuous Form 1422A Tyvek Autoclavable Tubing
Continuous Form tubing made from 1422A Tyvek® provides the user with tremendous sizing flexibility when wrapping pharmaceutical equipment for steam sterilization.
1422A Tyvek Filling Needle Bag
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. Also creates secondary level of protection during aseptic presentation.
Self-Sealing Medical Grade Sterilization Pouches
Designed for light duty, these economical sterilization pouches are designed for steam sterilization cycles (121 to 135 °C)
BHD Filling Needle Bags
These bags protect filling needles from accidental touching after they are removed from the primary sterilization packaging.
Self-Sealing BHD Autoclave Bags
The use of these 2 ply self-sealing bags will save valuable time in the equipment prep area. The 2-ply BHD material resists ripping and punctures.
BHD Autoclave Bags with Drawstring Closure
Standardize and simplify the way equipment is wrapped for steam sterilization.
Easy-Tear Tyvek Stopper Bags
Easy-Tear Feature for easier opening and sealing. No sterile cutting instruments required to open the bag
Self-Sealing 1073B Tyvek Chevron Peel Pouches
A significant upgrade over paper based steam sterilization pouches. These self-sealing Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do.
Heat Seal 1073B Tyvek Chevron Peel Pouches
These heat seal Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do
Continuous Form 1073B Tyvek Breather Bag Tubing
Unroll and cut to desired length, heat seal both ends. A significant upgrade over paper-based steam sterilization reels
Autoclavable at 121 to 125 °C
1073B Tyvek Stopper Bags
Stopper bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required.
1073B Tyvek Stopper Bag Tubing
Unroll and cut to desired length, heat seal both ends. Stopper bag tubing is ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
All 1073B Tyvek Heat Seal Autoclave Bags
100% Medical Grade 1073B Tyvek® Autoclave Bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
Self-Sealing, All 1073B Tyvek Autoclave Bags
Self-sealing closure reduces prep time and eliminates the need for and the validation of a heat sealing machine.
Self-Sealing Heavy Duty SMS Autoclave Bags
Designed for higher temperature steam sterilization cycles (135 °C)* and compatible with ethylene oxide and hydrogen peroxide sterilization.
1421B PureClean Tyvek Heat Seal Autoclave Bag
PureClean™ Autoclave bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
Self-Sealing 1421B PureClean Tyvek Autoclave Bag
Self-sealing PureClean™ Autoclave bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
1421B PureClean Tyvek Drawstring Autoclave Bag
PureClean™ Autoclave bags with drawstring closures are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
Continuous Form 1421B PureClean Tyvek Autoclavable Tubing
Continuous Form PureClean™ Tyvek Tubing is made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process..
1421B PureClean Tyvek Filling Needle Bag
PureClean™ Tyvek Filling Needle bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.