Ovens & Incubators
VWR® Ovens and Incubators provide superior temperature accuracy and excellent recovery time for a wide range of demanding applications. Forced air mechanical convection units offer uniform heating, precise temperature control, and fast processing using a blower that circulates heated air in a horizontal airflow pattern. Gravity convection units feature a vertical airflow pattern perfect for drying samples that require a low-turbulence environment.
Digital advanced and adaptive microprocessor control systems provide superior temperature accuracy. All units feature a durable, powder-coated steel exterior with stainless steel interior; glass wool insulation; and high-temperature grade, foamed silicone rubber door gasket.
Product selection
Carbon Dioxide Incubators
Ideal for busy laboratories where contamination concerns and ease of cleaning are important factors...
Fan Assisted and Natural Convection Ovens
Dry and heat your samples with precision. Browse our selection of forced air and gravity convection ovens...
Forced Air and Gravity Convection Incubators
Provide a stable environment for your samples with our selection of forced air and gravity incubators...
Low-Temperature / B.O.D. Incubators
Offer excellent temperature uniformity for a wide range of applications...
Vacuum Ovens
Suitable for drying, baking, conditioning, out-gassing solids and liquids, vacuum embedding, moisture testing, and age testing...