Equipment Covers
1422A Tyvek Mini-Muff Closures
Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size., untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications.
1422A Tyvek Mini-Muff Closures with Drawstring Closure
Each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
BHD Autoclavable Mini-Muffs
Autoclavable up to 135 °C, BHD Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses.
BHD Mini-Muff Closures with Tyvek® Drawstring Closure
Autoclavable up to 135 °C, each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
Blue Super-Duty Autoclavable SMS Muff Covers
Designed for autoclave cycles (121 to 135 °C, Super Duty SMS Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses.
Blue Heavy-Duty Autoclavable SMS Muff Covers
Designed for autoclave cycles (121 to 135 °C, Heavy Duty SMS Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses.
DuPont™ 1422A Tyvek® Mini-Muff Closures with Skirt
The skirt extension on these Muffs provides a safe area to remove the it without inadvertently touching the critical surfaces with the fingers.
DuPont™ 1422A Tyvek® Autoclavable Stopper Bowl Covers
Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
BHD Stopper Bowl Covers
Autoclavable up to 135 °C, leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
Super-Duty 88 GSM SMS Autoclavable Bowl Covers
Designed for autoclave cycles (121 to 135 °C) leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
Heavy-Duty 50 GSM SMS Autoclavable Bowl Covers
Designed for autoclave cycles (121 to 135 °C) leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
UV Light Blocking GMP Equipment Covers
Keeps biopharmaceutical pharmaceutical production equipment clean and protected from potentially damaging UV light.
GMP Covers, 1422A Tyvek®, Autoclavable, Cylindrical Shaped
Helps minimize microbial and particulate contamination risks in pharmaceutical production operations.
GMP Covers, 1422A Tyvek®
GMP Covers are available with elastic, zippers, snaps and see through windows and Identification Pouches
Polypropylene Mini-Muff Closures
Polypropylene muffs are a great choice in non-sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Rectangular Shaped DuPont™ Tyvek® 1422A Elasticized Covers for Steam Sterilization
Save time and standardize the activities in equipment preparation for sterilization. Leave the covers in place during installation to prevent microbial and particulate contamination.
1421B PureClean™ Tyvek Mini-Muff Closures
PureClean™ Muffs are made with a new untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.
PureClean™ 1421B Tyvek® Autoclavable Stopper Bowl Covers
Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.