Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
M00652 M00653 M00654 M00655 M00656 M00657 M00658 M00659 M00660 M00661 M00662 M00663 M00664 M00665 M00666 M00667 M00668 M00669 M41210 M41212 M41215 M42010 M42012 M42015
76510-572EA 104.84 USD
76510-572 76510-574 76510-576 76510-578 76510-580 76510-582 76510-584 76510-586 76510-588 76510-590 76510-592 76510-594 76510-596 76510-598 76510-600 76510-602 76510-604 76510-606 76510-626 76510-628 76510-630 76510-636 76510-638 76510-648 76510-650 76510-652 76510-654 76510-656 76510-658 76510-660 76510-662 76510-664 76510-666 76510-668 76510-670 76510-672
Bis-Tris Mini Protein Gels, GenScript®
Electrophoresis Gels Precast Gels
GenScript's Bis-Tris gels are high-performing polyacrylamide gels designed to separate a wide range of protein sizes by electrophoresis. The gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis and increases gel stability. Furthermore, the neutral pH which minimizes protein modification when compared to Tris-glycine gels.
  • Large well volume
  • High resoluton and reproducibility
  • Long shelf life
  • Cost effective
  • Compatible cassette design
SurePAGE™ gels are premium, high resolution with enhance reproducibility gels, while theMore Product Information