Slagging Hood
Slagging hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. These slagging hoods have a convenient steel post with a steel pad to hammer your lead buttons to remove the slag. There is also a chute where the slag is deposited for disposal. Designed for industrial and mining applications, our slagging hoods have a 10” flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
Slagging hoods are made of steel and blue powder coated for a long-lasting finish (additional colours available on request). Each unit comes complete with LED lighting and a compressed air hose. Standard size is 6ft but custom sizes are also available.