Cupellation Hood
Cupellation hood is designed for the MAS3 Furnace and is to be used in conjunction with a Furnace Cap Hood (CA11022-166). The cupellation hood bolts onto the front of the furnace cap hood and effectively collects lead fumes generated by the hot cupels after cupellation.
Cupellation hoods are made of steel and white powder coated for a long-lasting finish (additional colours available on request).