Intuvo Supplies
Keep your Intuvo GC running smoothly by regularly replacing your Intuvo consumables, including Intuvo gaskets, guard chips, flow chips, and jumper chips. Not sure what you'll need? Try the Agilent Intuvo Install kit.
Intuvo Flow Chips
Intuvo Flow Chips are modular, microfluidic components that create connections between the inlet, column, and detector(s) - easily swapped in minutes.
Intuvo Gas Clean Filter Kit
This gas clean filter system delivers clean gases, reducing the risks of column damage, sensitivity loss, and instrument downtime.
Intuvo Gaskets
The Agilent Intuvo GC system uses ferrule-free face seals for all fittings within the sample gas flow path.
Intuvo Guard Chips & Jumper Chips
The Agilent Intuvo 9000 GC system uses ferrule-free face seals for all fittings within the sample gas-flow path.
Intuvo Install Kit
This gas clean filter system delivers clean gases, reducing the risks of column damage, sensitivity loss, and instrument downtime.
Intuvo Tools
The Intuvo 9000 GC system uses a unique torque driver to secure the flow chip and guard chip.