Pathogen Detection with the BAX® System Q7
The Hygiena™ BAX® System offers advanced, automated detection of foodborne pathogens, spoilage organisms and other microbes in raw ingredients, finished products and environmental samples.
The BAX System uses the power of polymerase chain reaction (PCR) technology to automatically amplify and detect targeted bacterial DNA. First, the DNA is amplified through a series of timed heating and cooling cycles. These cycles produce an exponential increase in the number of targeted DNA fragments, if present, creating millions of copies in a very short time. If the target sequence is present, the BAX System automatically detects the amplified fragments by measuring a fluorescent signal within the sample. If the target sequence is not present, no detectable amplification takes place.
The BAX System Q7 instrument uses optimized PCR assays (sold separately) that target specific DNA fragments unique to each target organism, providing a stable, reliable indicator that the organism is present and vastly reducing the potential for cross reactions with similar organisms. To further simplify the process, the BAX System assays provide all requisite PCR primers, polymerase and nucleotides as a stable, dry, manufactured tablet in sealed PCR tubes to eliminate the need for reagent preparation. After amplification, these tubes remain sealed for the detection phase, significantly reducing the potential for sample or laboratory contamination with the amplified PCR product.
Product selection
Salmonella Testing
Validated real-time and standard PCR assays available for detecting Salmonella species in a variety of food and environmental samples
Listeria Testing
Validated real-time and standard PCR assays available for detecting Listeria species and L. monocytogenes in a variety of food and environmental samples
E. coli O157:H7 Testing
Validated real-time and standard PCR assays available for detecting E. coli O157:H7 in produce, meats and other samples
E. coli STEC Testing
Validated real-time PCR assays for detecting stx/eae and the "top six" non-O157:H7 STEC in ground beef and beef trim
Staphylococcus Testing
Validated real-time PCR assay for detecting Staphylococcus aureus in beef, soy protein and infant formula
Vibrio Testing
Validated real-time PCR assay for detecting V. cholera, V. parahaemolyticus and V. vulnificus in seafoods
Cronobacter Testing
Standard PCR assay for detecting Cronobacter species (E. sakazakii) in dried dairy, soy and infant formula and environmental samples
Yeast & Mold Testing
Validated standard PCR assay for detecting yeasts and molds
Campylobacter Testing
Real-time PCR assay for detecting Campylobacter jejuni, coli and lari in poultry and poultry rinse samples
Shigella Testing
Real-time PCR assay designed to quickly eliminate negative results, reducing the number of samples that must be confirmed.