Hygiena™ BAX® System – Fast, Accurate, Easy to Use Pathogen Detection
The Hygiena™ BAX® System offers advanced, automated detection of foodborne pathogens, spoilage organisms and other microbes in raw ingredients, finished products and environmental samples. As part of your quality control system, the automated DNA-based test provides rapid results to help you make product release decisions with confidence, protecting your company and your brand.
The BAX® System Q7 instrument produces fast, accurate, simple results for up to 96 samples at a time for customers needing higher sample throughput.
The BAX® System X5 instrument provides the same advanced testing capabilities for customers who prefer greater testing flexibility, offering a smaller, lightweight construction that processes up to 32 samples per batch.
Pathogen Detection with the BAX® System Q7
The Hygiena™ BAX® System offers advanced, automated detection of foodborne pathogens, spoilage organisms and other microbes in raw ingredients, finished products and environmental samples...
Pathogen Detection with the BAX® System X5
The BAX® System X5 offers the same fast, accurate and easy-to-use pathogen detection solution in a smaller, lightweight construction to provide greater testing flexibility...