Dithiothreitol (DTT, Cleland's reagent)

Supplier: VWR
Warning

M109-25G M109-5G
97063-758CS 6890.67 USD
97063-758 97063-760
Dithiothreitol (DTT, Cleland's reagent)
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
Melting Pt: 42 °C…43 °C
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=aebd7f10ad67505739d64fa5ba9b05c53205beea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@283nm (0.02M, Water) <= 0.05
DNase none detected
Electrophoresis (P/F) Pass
IR Pass
Loss on Drying <= 0.5%
Melting Point 39- 43°C
Oxidized DTT <= 0.5%
Protease none detected
Purity (-SH Content) >= 99.4%
RNase none detected
Solubility (5%, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR