EasySep™ Human Memory CD4+ T Cell Enrichment Kits
Supplier: STEMCELL Technologies
19157 19157RF
MSPP-19157EA 1429.23 USD
MSPP-19157 MSPP-19157RF
EasySep™ Human Memory CD4+ T Cell Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human memory CD4+ T cells
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Untouched, viable cells
The EasySep™ Human Memory CD4 T Cell Enrichment Kit is designed to isolate memory CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8, CD14, CD16, CD19, CD20, CD36, CD45RA, CD56, CD123, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Precaución: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').