RosetteSep™ Human CD4+ T Cell Enrichment Cocktails
Supplier: STEMCELL Technologies
15062 15022
MSPP-015062EA 1473.85 USD
MSPP-015062 MSPP-15022
RosetteSep™ Human CD4+ T Cell Enrichment Cocktails
Cell Separation Media
Immunodensity isolation of untouched CD4+ T cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Isolated cells are untouched
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD4+ T Cell Enrichment Cocktail is designed to isolate CD4+ T cells from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (STMC15705) or Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified CD4+ T cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Precaución: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).