Puromycin dihydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

A547954-100MG A547954-1MG A547954-250MG A547954-50MG
77602-314EA 194.75 USD
77602-314 77669-414 77669-416 77669-418
Puromycin dihydrochloride ≥98%
Puromycin dihydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
MDL Number: MFCD00012691
CAS Number: 58-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR