CloneR™ Serum Free Suplement
Supplier: STEMCELL Technologies
05889 05888
MSPP-05889PK 560 USD
MSPP-05889 MSPP-05888
CloneR™ Serum Free Suplement
Cell Culture Media
Defined supplement for single-cell cloning of human ES and iPS cells.
CloneR™ is a defined, serum-free supplement designed to increase the cloning efficiency and single-cell survival of human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPS cells). CloneR™ enables the robust generation of clonal cell lines without single-cell adaptation, thus minimizing the risk of acquiring genetic abnormalities. CloneR™ is compatible with the TeSR™ family of media for human ES and iPS cell maintenance as well as your choice of cell culture matrix. For improved single-cell survival in additional applications, and higher cloning efficiency, try the CloneR™2 supplement.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').