Phorbol-12-myristate-13-acetate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
TPA, Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a-Acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate
A175370-5MG
A175370-50MG
A175370-100MG
A175370-10MG
77567-588EA
143.17
USD
77567-588
77600-420
77667-442
77667-444
Phorbol-12-myristate-13-acetate 95%
Phorbol-12-myristate-13-acetate
Precaución: For research use only.
Formula:
C₃₆H₅₆O₈ MW: 616.836 g/mol |
MDL Number:
MFCD00036736 CAS Number: 16561-29-8 UN: 2928 ADR: 6.1,8,Ⅰ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...