Anti-SARS-CoV-2 RBD Rabbit Monoclonal Antibody [clone: 4G6]

Supplier: Genscript
A02053-1 A02053-100
76510-138EA 3197.7 USD
76510-138 76510-140
Anti-SARS-CoV-2 RBD Rabbit Monoclonal Antibody [clone: 4G6]
Antibodies
The product is specific for SARS-CoV-2 Spike Protein S1 subunit and its RBD domain.

  • COVID-19 antibodies
  • Validated for ELISA, WB, FACS, and SARS-CoV-2 virus neutralization assay
  • Control antibodies and various antibody pairs available for ELISA-based detection
  • Neutralizing Antibody Standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples

Genscript offers site-specific Neutralizing antibodies, neutralizing antibody standards, and control antibodies.

Type: Primary
Antigen: SARS-CoV-2 RBD
Clonality: Monoclonal
Clone: 4G6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR