Dithiothreitol (DTT, Cleland's reagent) 95%

Supplier: Ambeed
Danger

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

A587635-10G A587635-1G A587635-1MG A587635-25G A587635-5G A587635-100G A587635-500G
76805-860EA 13 USD
76805-860 76805-862 76805-864 76805-866 76805-868 77551-768 77572-870
Dithiothreitol (DTT, Cleland's reagent) 95%
Dithiothreitol (DTT, Cleland's reagent)

Precaución: for Research Use Only

Formula: C₄H₁₀O₂S₂
MDL Number: MFCD00004877
CAS Number: 3483-12-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR