Phorbol-12-myristate-13-acetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TPA, Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a-Acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate

35615-0010 35615-0050
TS35615-0010EA 104 USD
TS35615-0010 TS35615-0050
Phorbol-12-myristate-13-acetate 97%
Phorbol-12-myristate-13-acetate
Formula: C₃₆H₅₆O₈
MW: 616.836 g/mol
Melting Pt: 72 °C
Storage Temperature: Freezer
MDL Number: MFCD00036736
CAS Number: 16561-29-8

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Film or foam
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR