Polyethylene glycol diacrylate (PEGDA), M.W. 3500, powder, not sterile

Supplier: ADVANCED BIOMATRIX, INC. MS

Synonyms: PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate

GS700 GS705
103792-968EA 287.69 USD
103792-968 103792-970
Polyethylene glycol diacrylate (PEGDA), M.W. 3500, powder, not sterile
Polyethylene glycol diacrylate (PEGDA)

Polyethylene Glycol Diacrylate (PEGDA) is a 3500 MW blank slate hydrogel that gels rapidly at room temperature in the presence of a photoinitiator and light. PEGDA is hydrophilic, elastic and can be customized to include a variety of biological molecules.


  • 3500 MW
  • Highly crosslinkable PEGDA
  • Optimal for "blank slate" 3D hydrogels


Polyethylene Glycol Diacrylate (PEGDA) hydrogels are powerful tools for uncovering basic cellular biology because they are considered biologically inert (“blank slate”) and their mechanical properties can be varied over a large range of moduli.

PEGDA is an emerging scaffold for tissue engineering and regenerative medicine since polymerization can occur rapidly at room temperature and requires low energy input, has high water content, is elastic, and can be customized to include a variety of biological molecules.

PEGDA is a crosslinker repeating polymer crosslinker and is >80% acrylated. It can be used as a thiol-reactive crosslinker in our HyStem systems at low concentration, or can be used to make a PEGDA-only hydrogel in the presence of a free-radical chain photoinitiator and light source. PEG-only gels do not typically support cell attachment without the incorporation of cellular attachment sites.


Precaución: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Formula: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Storage Temperature: Freezer
MDL Number: MFCD00081876
CAS Number: 26570-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR