Thiol-Modified Hyaluronan/Heparin Mixture, Heprasil®
Supplier: ADVANCED BIOMATRIX, INC. MS
Heprasil® is a mixture of thiol-modified hyaluronic acid thiol-modified heparin. Heprasil® is a component of the HyStem®-HP kits, but can be purchased separately here.
- Thiolated hyaluronic acid and heparin
- Ideal for sustained growth factor release
- Use in conjunction with Extralink for 3D hydrogels
Heprasil® is thiol-modified hyaluronic acid with thiol-modfied heparin and is a component of the HyStem® hydrogel kit. Hyaluronic acid is a major constituent of native extracellular matrix (ECM). Heparin is also present in the ECM as heparan sulfate. Most cells do not attach to Heprasil-only hydrogels. Heprasil must be used in conjunction with Gelin-S® or ECM proteins such as laminin, collagen, or fibronectin for most 3-D cell culture and tissue-engineering applications.
Precaución: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.