5-Carboxyfluorescein
Supplier: Anaspec Inc
Synonyms:
Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate
AS-81003
AS-81003-1
AS-81003-5
103010-754EA
119.85
USD
103010-754
103010-756
103010-758
5-Carboxyfluorescein
5-Carboxyfluorescein
5-FAM is a single isomer. It is one of the most popular green fluorescent reagents used for in situ labeling peptides, proteins and nucleotides. It has also been used to prepare various small fluorescent molecules.
- Spectral properties: Abs/Em = 492/518 nm
- Solvent system: DMF or DMSO
- Appearance: Orange solid
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: RT
Formula:
C₂₁H₁₂O₇.xH₂O MW: 376.32 g/mol (anhy) Storage Temperature: Freezer |
MDL Number:
MFCD00036874 CAS Number: 76823-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...