Biotin-X NTA

Supplier: Anaspec Inc

Synonyms: Biotin-X nitrilotriacetic acid, tripotassium salt

AS-60655
103003-304EA 220.69 USD
103003-304
Biotin-X NTA
Biotin-X NTA

Excellent reagent for detecting polyhistidine-containing biomolecules


  • Spectral properties: Abs/Em = NA/NA nm
  • Solvent system: water
  • Appearance: solid
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₂₆H₄₀K₃N₅O₉S
MW: 715.98 g/mol
Storage Temperature: Freezer
CAS Number: 856661-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR