proBDNF Rapid ELISA Kit: Human, Mouse, Rat, Biosensis®

Supplier: Biosensis

BEK-2237-1P BEK-2237-2P
76264-594EA 437.85 USD
76264-594 76264-596
proBDNF Rapid ELISA Kit: Human, Mouse, Rat, Biosensis®
Assays ELISAs

The Biosensis proBDNF RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the specific, fast and reliable quantification of proBDNF in less than 4 hours in human serum and plasma (citrate and EDTA), cell culture supernatants, mouse and rat cell lysates and rat brain only if used as directed.


Please refer to the kit protocol for specific use instructions for each substrate application, in particular human blood samples. This ELISA kit consists of a pre-coated monoclonal anti-proBDNF capture antibody, a biotinylated anti-matureBDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of proBDNF present in samples and protein standards.


A proBDNF positive control (QC sample) is provided to assure consistent assay performance. This proBDNF ELISA kit employs a recombinant, cleavage-resistant human proBDNF standard produced in mammalian cells by Biosensis and validated against externally available proBDNF proteins.


Note that accurate proBDNF quantification in human serum and EDTA-plasma requires the addition of Heterophilic Antibody Blocker BL-004-500 provided in the kit, and available for purchase separately. This ELISA kit has not been tested for other applications.


Información de suministro: The ELISA kit box contains 1 or 2×96-well pre-coated strip plate, protein standards, QC sample, detection reagents, heterophilic antibody blocker, wash and sample buffers, substrate buffer and detailed protocols.


Precaución: It has been configured for research use only and is not to be used for diagnostic or clinical procedures.

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein proBDNF proBDNF
Primary antibody reactivity Human Human
Assay Type (ELISA with LOV) Sandwich Sandwich
Host
Sensitivity Typical limit of detection (LOD) for proBDNF is 6 pg/ml determined as 150% of the blank value Typical limit of detection (LOD) for proBDNF is 6 pg/ml determined as 150% of the blank value
Detection Range 15.6 - 1000 pg/ml 15.6 - 1000 pg/ml
Cross Reactivity The capture antibody used in this ELISA kit binds to an epitope within the pro-domain of proBDNF. Thus, this ELISA detects the full length form of proBDNF and does not quantify mature BDNF. Whether this ELISA kit can detect truncated BDNF is unknown at present.
Mature BDNF cross-reactivity was assessed by spiking 28 kDa mature BDNF protein obtained from WHO into a 1/5 diluted human serum sample at 5 ng/ml, which represents a BDNF concentration level of 25 ng/ml in undiluted, normal human serum. Cross-reactivity of mature BDNF was <0.3% (w/v), or <0.1% in molar concentration. The assay antibodies do not cross-react with nerve growth factor (NGF), neurotrophin-3 (NT-3) or NT-4/5
The capture antibody used in this ELISA kit binds to an epitope within the pro-domain of proBDNF. Thus, this ELISA detects the full length form of proBDNF and does not quantify mature BDNF. Whether this ELISA kit can detect truncated BDNF is unknown at present.
Mature BDNF cross-reactivity was assessed by spiking 28 kDa mature BDNF protein obtained from WHO into a 1/5 diluted human serum sample at 5 ng/ml, which represents a BDNF concentration level of 25 ng/ml in undiluted, normal human serum. Cross-reactivity of mature BDNF was < 0 .3% (w/v), or < 0.1% in molar concentration.
The assay antibodies do not cross-react with nerve growth factor (NGF), neurotrophin-3 (NT-3) or NT-4/5
Specificity Human, mouse and rat Human, mouse and rat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR