HiLyte™ Fluor 488 acid, SE fluorescent dye
Supplier: Anaspec Inc
AS-81161
AS-81161-1
103010-874EA
760
USD
103010-874
103010-876
HiLyte™ Fluor 488 acid, SE fluorescent dye
HiLyte™ Fluor
HiLyte™ Fluor 488 acid, SE is an excellent amine-reactive fluorescent labeling dye that generates the protein conjugates far superior to those of fluorescein derivatives such as FITC.
- Spectral Properties Abs/Em = 502/527 nm
- Solvent System DMF or DMSO
- Storage -20°C desiccated and protected from light
Not only are HiLyte™ Fluor 488 conjugates significantly brighter than fluorescein conjugates, they are much more photostable. Additionally the fluorescence of HiLyte™ Fluor 488 fluorophore is independent of pH from 4 to 10. This pH insensitivity is a major improvement over fluorescein, which emits its maximum fluorescence only above pH 10.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...