Wasp Mastoparan
Supplier: Anaspec Inc
This 14-residue peptide toxin from the wasp venom is originally found as a histamine releaser from mast cells. It induces mitochondrial membrane permeabilization via a CsA-inhibitable mechanism.
Sequence:INLKALAALAKKIL-NH2
MW:1478.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...