PKG Substrate
Supplier: Anaspec Inc
AS-60298-1
AS-60298-5
103003-092EA
165.15
USD
103003-092
103003-094
PKG Substrate
Proteins and Peptides
RKRSRAE is a selective substrate for protein kinase G (PKG) with a strong preference for PKG Iα (Km = 59 µM) over PKG II (Km = 305 µM).
Sequence:RKRSRAE
MW:902 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...