Kemptide, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
Kemptide is a phosphate acceptor peptide that serves as a synthetic substrate for PKA (Km = 16 µM). The corresponding fluorescent and biotinylated peptides are also proven to be good substrates for PKA.
Sequence:5-FAM-LRRASLG
MW:1130.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...