GRGDSP Peptide
Supplier: Anaspec Inc
AS-22945
AS-22946
102996-334EA
61.38
USD
102996-334
102996-336
GRGDSP Peptide
Proteins and Peptides
An inhibitor of cell attachment to salmosin and vitronectin.
Sequence:GRGDSP
MW:587.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...