Dynorphin A (1-17)
Supplier: Anaspec Inc
Dynorphins are a class of opioid peptides that arise from the precursor protein prodynorphin. Upon cleavage by proprotein convertase 2 (PC2), multiple active peptides are released: dynorphin A, dynorphin B, and α/β-neo-endorphin. Dynorphins exert their effects primarily through the κ-opioid receptor (KOR), a G-protein-coupled receptor. Dynorphin has been shown to be a modulator of pain response, involved in drug addiction and appetite control.
Sequence:YGGFLRRIRPKLKWDNQ
MW:2147.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...