HIV TAT (47-57) FITC LC Peptide, FITC (Fluorescein Isothiocyanate)
Supplier: Anaspec Inc
AS-27042
AS-27043
103000-576EA
289.38
USD
103000-576
103000-578
HIV TAT (47-57) FITC LC Peptide, FITC (Fluorescein Isothiocyanate)
Proteins and Peptides
This fluorescent (FITC)-labeled TAT peptide contains a long chain (LC) to prevent FITC from degradation. Abs/Em = 493/522 nm.
Sequence: FITC-LC-YGRKKRRQRRR-NH2
MW: 2061.4 Da
% Peak area by HPLC: 95
Storage condition: -20°C
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...