Human Ang II Peptide

Supplier: Anaspec Inc

AS-20633 AS-20634
102996-066EA 92.08 USD
102996-066 102996-068
Human Ang II Peptide
Proteins and Peptides

The octapeptide angiotensin II (Ang II) exerts a wide range of effects on the cardiovascular system. It is also implicated in the regulation of cell proliferation, fibrosis and apoptosis. Ang II is formed by cleavage of Ang I by the angiotensin-converting enzyme (ACE) or chymases. Human heart chymase, a chymotrypsin-like serine proteinase, hydrolyzes the Phe8-His9 bond to yield the octapeptide hormone angiotensin II and His-Leu.
Sequence: DRVYIHPF
MW: 1046.2 Da
% Peak area by HPLC: 95
Storage condition: -20°C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR