Human;Mouse;Rat Beta-Amyloid (17-40)
Supplier: Anaspec Inc
AS-22813- AS-22814
102999-340EA 279.15 USD
102999-340 102999-342
Human;Mouse;Rat Beta-Amyloid (17-40)
Proteins and Peptides
In Alheimer's Disease brains, plaques contain amino-terminal truncated beta-Amyloid peptides including the alpha secretase-generated p3 fragments, beta-Amyloid 17-40 and 17-42. They were shown to induce pro-inflammatory cytokine and chemokine production in vitro and in vivo.
  • Peptide Content: ≥ 60%