Human Beta-Amyloid (5-42)
Supplier: Anaspec Inc
AS-60087-01
AS-60087-1
103002-974EA
279.15
USD
103002-974
103002-976
Human Beta-Amyloid (5-42)
Proteins and Peptides
This peptide is beta-amyloid N-terminally truncated to obtain 5-42 peptide.
Sequence: RHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
Molecular Weight: 4051.6 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...