2-Methyl-5-nitroimidazole-1-acetic acid (synthetic) ≥98% (by GC)
Supplier: ADIPOGEN CORP MS
CDX-M0091-M100
CDX-M0091-G001
102990-476EA
210.46
USD
102990-476
102990-478
2-Methyl-5-nitroimidazole-1-acetic acid (synthetic) ≥98% (by GC)
2-Methyl-5-nitroimidazole-1-acetic acid
2-Methyl-5-nitroimidazole-1-acetic acid is a main acid metabolite of the antibiotic metronidazole.
- Can be used as analytical reference material
- Main acid metabolite of metronidazole
- Shows anti-bacterial activity
A main acid metabolite of the antibiotic metronidazole showing some anti-bacterial activity. Compound can be used as analytical reference material.
White to light yellow/beige powder
Soluble in chloroform
Storage Temperature:
Freezer |
CAS Number:
1010-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...