Josamycin (synthetic) ≥98% (by UV method)

Supplier: ADIPOGEN CORP MS

Synonyms: (2S,3S,4R,6S)-6-(((2R,3S,4R,5R,6S)-6-(((4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-Acetoxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)oxacyclohexadeca-11,13-dien-6-yl)oxy)-4-(dimethylamino)-5-hydroxy-2-methyltetrahydro-2H-pyran-3-yl)oxy)-4-hydroxy-2,4-

CDX-J0001-M050 CDX-J0001-M200 CDX-J0001-GG25
102990-308EA 66.46 USD
102990-308 102990-310 102990-312
Josamycin (synthetic) ≥98% (by UV method)
Josamycin

Josamycin is a member of the leucomycin family of macrolide antibiotics.


  • Anti-microbial against a wide variety of pathogens
  • Macrolide antibiotics
  • Produced by Streptomyces kitasatoensis


A member of the leucomycin family of macrolide antibiotics produced by Streptomyces kitasatoensis. May overcome anti-cancer drug resistance by inhibiting the binding of vinblastine or cyclosporin A to P-glycoprotein (Pgp). An anti-microbial against a wide variety of pathogens.


White to off-white powder


Soluble in water

Formula: C₄₂H₆₉NO₁₅
MW: 827.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00211043
CAS Number: 16846-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR