Clarithromycin N-oxide (synthetic) ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
CDX-C0310-M010
CDX-C0310-M025
CDX-C0310-M100
102989-582EA
105.23
USD
102989-582
102989-584
102989-586
Clarithromycin N-oxide (synthetic) ≥98% (by HPLC)
Clarithromycin N-oxide
Clarithromycin N-oxide can be used as analytical reference material.
- Biologically inactive, urinary metabolite
- Analytical reference material
Biologically inactive urinary metabolite of Clarithromycin (oxidized by ozone), a macrolide antibiotic. Compound can be used as analytical reference material.
White powder
Soluble in methanol and chloroform
Storage Temperature:
Freezer |
CAS Number:
118074-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...