6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) (synthetic) ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 6-Carboxytetramethylrhodamine N-succinimidyl ester, 6-TAMRA, SE, 6-TAMRA N-succinimidyl ester

CDX-C0058-M005 CDX-C0058-M025
102987-652EA 290.77 USD
102987-652 102989-438
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) (synthetic) ≥97% (by HPLC)
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

6-TAMRA N-succinimidyl ester is an amine-reactive form of 6-carboxytetramethylrhodamine.


  • Used for labeling of peptides
  • Automated DNA sequencing
  • Labels aminoglycoside antibiotics


Amine-reactive form of 6-carboxytetramethylrhodamine single isomer used for labeling of peptides and aminoglycoside antibiotics. It is also most commonly used in automated DNA sequencing.


Colour: Dark red


Soluble in DMSO or DMF

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Storage Temperature: Freezer
CAS Number: 150810-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR