Pentabromopseudilin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0441-C500 BVT-0441-M001
102989-026EA 221.54 USD
102989-026 102989-028
Pentabromopseudilin ≥98% (by HPLC)
Pentabromopseudilin

Pentabromopseudilin is an anti-bacterial, cytotoxic, herbicidal and anti-malarial compound.


  • Antibiotic
  • Potent myosin ATPase inhibitor
  • Anti-bacterial


Antibiotic. Human lipoxygenase inhibitor. Potent myosin ATPase inhibitor. Inhibitor of the non-mevalonate pathway enzyme IspD.


Off-white powder


Soluble in DMSO (5mg/ml), acetone, butanol, ethylacetate or diethylether

MW: 553.7 g/mol
Storage Temperature: Freezer
CAS Number: 10245-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR