Cephalochromin ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Cephalochromin inhibits NO production by activated macrophages.
- Anti-tumor and cytotoxic compound
- Mycotoxin
- Antibiotic
Mycotoxin. Bis(naphtha-gamma-pyrone) derivative similar to chaetochromin and ustilaginoidin. Antibiotic. Anti-bacterial, anti-fungal and anti-algal compound. Calmodulin-sensitive PDE activity inhibitor. Selective Fabl (bacterial enoyl-acyl carrier protein (ACP) reductase) inhibitor. Inducer of G0/G1 cell cycle arrest and apoptosis in human A549 cells.
Orange Amorphous powder
Soluble in DMSO, methanol, acetone or dichloromethane
MW:
518.5 g/mol Storage Temperature: Freezer |
CAS Number:
25908-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...