Helvolic acid ≥96% (by HPLC, NMR)
Supplier: ADIPOGEN CORP MS
Helvolic acid is a steroidal triterpene antibiotic.
- Anti-bacterial
- Mycotoxin
- Oxidized LDL uptake inhibitor
Anti-bacterial. Shows broad activity against Gram positive and negative bacteria. Mycotoxin. Antifungal. Phytotoxin. Inhibitor of the protein biosynthesis through archeal elongation factor 2 (EF-2).
Off-white solid
Soluble in DMSO, acetone or chloroform
Storage Temperature:
Freezer |
CAS Number:
29400-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...