3,28-Di-O-(3,3-dimethylglutaryl)betulin ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
BVT-0404-M001
BVT-0404-M005
102981-210EA
131.54
USD
102981-210
102981-212
3,28-Di-O-(3,3-dimethylglutaryl)betulin ≥97% (by HPLC)
3,28-Di-O-(3,3-dimethylglutaryl)betulin
Semi-synthetic derivative of betulin (from birch bark). Potent antiviral compound. Shows anti-HIV activity (more active than AZT in vitro).
Specification Test Results
Source/Host | Semi-synthetic derivative from betulin (from birch bark). |
Purity |
|
Appearance/Color | White to off-white solid. This substance is hygroscopic and might gradually change its state of aggregation from solid to oil. |
Solubility | Soluble in methanol, acetone or DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...