GV-58 ≥97% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
(R)-2-((6-(((5-Methylthiophen-2-yl)methyl)amino)-9-propyl-9H-purin-2-yl)amino)butan-1-ol, (2R)-2-[[6-[[(5-Methyl-2-thienyl)methyl]amino]-9-propyl-9H-purin-2-yl]amino]-1-butanol
AG-MR-C0035-M001
AG-MR-C0035-M005
102981-190EA
83.08
USD
102981-190
102981-192
GV-58 ≥97% (by NMR)
GV-58
Selective N- and P/Q-type Ca2+-channel agonist. These Ca2+-channels regulate transmitter release in synapses. Potential lead compound for a variety of disorders that result in neuromuscular weakness.
Formula:
C18H26N6OS MW: 374.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD28385865 CAS Number: 1402821-41-3 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...