Smoothened agonist dihydrochloride hydrate ≥98% (by HPLC), water soluble

Supplier: ADIPOGEN CORP MS

Synonyms: SAG hydrochloride hydrate

AG-CR1-3585-M001 AG-CR1-3585-M005
102988-654EA 69.23 USD
102988-654 102988-656
Smoothened agonist dihydrochloride hydrate ≥98% (by HPLC), water soluble
Smoothened agonist dihydrochloride hydrate

Cell permeable Sonic hedgehog (Shh) agonist. Cell permeable smoothened (Smo) agonist. Induces Sonic hedgehog (Shh) pathway activation and counteracts cyclopamine inhibition of Smo. Acts as an activator of Smo at low concentrations and as an inhibitor of Smo at very high concentrations. Potent activator of VEGF expression in vitro.

Formula: C₂₈H₂₈ClN₃OS·2HCl·3H₂O
MW: 617 g/mol
Storage Temperature: Freezer
CAS Number: 364590-63-6

Order Now

Specification Test Results

Purity 98% (HPLC).
Appearance/Color Light yellow solid.
Solubility Soluble in water or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR