SB203580 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: SB 203580, 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine

SYN-1074-M001 SYN-1074-M005 SYN-1074-M010 SYN-1074-M050
102991-840EA 74.77 USD
102991-840 102991-842 102991-844 102991-846
SB203580 ≥95%
SB203580

SB203580 is a specific, ATP competitive, cell-permeable inhibitor of p38 MAPK with IC(50) of 0.3-0.5µM. It is 10-fold less sensitive to SAPK3(106T) and SAPK4(106T) and blocks PKB phosphorylation with IC(50) of 3-5µM.

Formula: C₂₁H₁₆FN₃OS
MW: 377.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00922198
CAS Number: 152121-47-6

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR