SB202190 ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
4-(4-(4-Fluorophenyl)-5-(pyridin-4-yl)-1H-imidazol-2-yl)phenol, SB 202190
SYN-1073-M001
SYN-1073-M005
SYN-1073-M010
SYN-1073-M050
102991-824EA
74.77
USD
102991-824
102991-826
102991-828
102991-830
SB202190 ≥95%
SB202190
SB202190 is potent and selective inhibitor of the p38alpha and beta, with IC(50) values of 50 and 100nM respectively. It is a powerful regulator of LPS-induced mRNAs in monocytes.
Formula:
C₂₀H₁₄FN₃O MW: 331.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00941964 CAS Number: 152121-30-7 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...