PD 184,352 ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(2-Chloro-4-iodoanilino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, 2-((2-Chloro-4-iodophenyl)amino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, PD 184352, CI-1040

SYN-1031-M001 SYN-1031-M005 SYN-1031-M010 SYN-1031-M050
102991-486EA 74.77 USD
102991-486 102991-488 102991-490 102991-492
PD 184,352 ≥95%
PD 184,352

CI-1040 (PD184352) is an orally active, highly specific, small-molecule inhibitor of MEK1/2, which is a key component of the ERK/MAPK pathway. It has an IC(50) of 17nM.

Formula: C₁₇H₁₄ClF₂IN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD02683961
CAS Number: 212631-79-3

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR