Vindoline ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-V0002-M050 CDX-V0002-M250 CDX-V0002-GG25
102991-160EA 105.23 USD
102991-160 102991-162 102991-164
Vindoline ≥98% (by HPLC)
Vindoline

Reference standard.

Formula: C₂₅H₃₂N₂O₆
MW: 456.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27991347
CAS Number: 2182-14-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR